Assay Details
Target Gene Details
Entrez Gene ID: | 23516 |
Gene Name: | solute carrier family 39 member 14 |
Gene Aliases: |
HMNDYT2, LZT-Hs4, NET34, ZIP14, cig19 |
Location: |
Chr.8:22367249-22434129 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC39A14 | NM_001128431.2 | NP_001121903.1 | ||
NM_001135153.1 | NP_001128625.1 | |||
NM_001135154.1 | NP_001128626.1 | |||
NM_015359.4 | NP_056174.2 | |||
XM_005273465.2 | XP_005273522.1 | |||
XM_006716323.2 | XP_006716386.1 | |||
XM_006716324.2 | XP_006716387.1 | |||
XM_011544478.2 | XP_011542780.1 | |||
XM_017013292.1 | XP_016868781.1 | |||
XM_017013293.1 | XP_016868782.1 | |||
XM_017013294.1 | XP_016868783.1 | |||
AK172810.1 | BAD18780.1 | |||
AK295807.1 | ||||
BC015770.1 | AAH15770.1 | |||
D31887.1 | ||||
DC323208.1 | ||||
DC423882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6116 | Chr.8:22368575 - 22399366 on Build GRCh38 | Insertion | SLC39A14 |
nsv831266 | Chr.8:22315953 - 22476033 on Build GRCh38 | Loss | SLC39A14 PIWIL2 PPP3CC |
nsv518646 | Chr.8:22296620 - 22520605 on Build GRCh38 | Gain | SLC39A14 PIWIL2 PPP3CC |
esv2764071 | Chr.8:22342635 - 22519221 on Build GRCh38 | Gain | SLC39A14 PIWIL2 PPP3CC |
esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss | C8orf58 GFRA2 POLR3D LOC101929172 EGR3 XPO7 BIN3-IT1 PEBP4 SLC39A14 LOC286114 TNFRSF10D PDLIM2 LOC389641 BIN3 DMTN HR LOC107984124 MIR320A BMP1 SFTPC LOC107986876 RHOBTB2 LGI3 FGF17 NPM2 TNFRSF10A LOC101929237 CHMP7 PIWIL2 LOC286059 NUDT18 TNFRSF10B SORBS3 TNFRSF10C DOK2 FAM160B2 LOC100507071 LOC102467222 PHYHIP REEP4 CCAR2 PPP3CC LOC254896 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |