Assay Details
Target Gene Details
Entrez Gene ID: | 100130742 |
Gene Name: | leucine rich repeat containing 69 |
Gene Aliases: |
- |
Location: |
Chr.8:91102619-91219236 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC69 | NM_001129890.1 | NP_001123362.1 | ||
AK097393.1 | ||||
AK130865.1 | BAC85448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2656822 | Chr.8:91103277 - 91173399 on Build GRCh38 | Deletion | LRRC69 |
esv3572901 | Chr.8:91101144 - 91173507 on Build GRCh38 | Loss | LRRC69 |
nsv611721 | Chr.8:91094842 - 91171430 on Build GRCh38 | Loss | LRRC69 |
nsv831390 | Chr.8:91024783 - 91186377 on Build GRCh38 | Gain | LRRC69 OTUD6B TMEM55A OTUD6B-AS1 |
esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss | LRRC69 OTUD6B TMEM55A C8orf88 SLC26A7 MIR4661 OTUD6B-AS1 |
esv3618014 | Chr.8:91103260 - 91173374 on Build GRCh38 | Loss | LRRC69 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |