Assay Details
Target Gene Details
Entrez Gene ID: | 5820 |
Gene Name: | Pvt1 oncogene (non-protein coding) |
Gene Aliases: |
LINC00079, MYC, NCRNA00079, onco-lncRNA-100 |
Location: |
Chr.8:127794533-128101253 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PVT1 | NR_003367.3 | |||
BM821521.1 | ||||
BX280136.1 | ||||
M25803.1 | ||||
M34428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831458 | Chr.8:127702603 - 127875740 on Build GRCh38 | Loss | MYC MIR1204 PVT1 CASC11 |
esv2752208 | Chr.8:127632573 - 127895715 on Build GRCh38 | Gain | MYC MIR1204 PVT1 CASC11 |