Assay Details
Target Gene Details
Entrez Gene ID: | 137964 |
Gene Name: | glycerol-3-phosphate acyltransferase 4 |
Gene Aliases: |
1-AGPAT 6, AGPAT6, LPAAT-zeta, LPAATZ, TSARG7 |
Location: |
Chr.8:41578188-41625001 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPAT4 | NM_178819.3 | NP_848934.1 | ||
XM_011544392.2 | XP_011542694.1 | |||
XM_011544393.1 | XP_011542695.1 | |||
XM_011544394.1 | XP_011542696.1 | |||
XM_017013042.1 | XP_016868531.1 | |||
XM_017013043.1 | XP_016868532.1 | |||
AF317516.1 | AAG33063.1 | |||
AF406612.1 | AAP21893.1 | |||
AY358670.1 | AAQ89033.1 | |||
AY513610.1 | AAS82774.1 | |||
BC051377.1 | AAH51377.2 | |||
BC061884.1 | AAH61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831299 | Chr.8:41448617 - 41692486 on Build GRCh38 | Loss | MIR486-1 ANK1 LOC102723729 GPAT4 GOLGA7 MIR486-2 NKX6-3 GINS4 |
esv3576565 | Chr.8:41064045 - 41790840 on Build GRCh38 | Gain | MIR486-1 SFRP1 ANK1 LOC102723729 GPAT4 GOLGA7 MIR486-2 NKX6-3 MIR548AO GINS4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |