Assay Details
Target Gene Details
Entrez Gene ID: | 6575 |
Gene Name: | solute carrier family 20 member 2 |
Gene Aliases: |
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1 |
Location: |
Chr.8:42416462-42542213 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC20A2 | NM_001257180.1 | NP_001244109.1 | ||
NM_001257181.1 | NP_001244110.1 | |||
NM_006749.4 | NP_006740.1 | |||
XM_005273613.3 | XP_005273670.1 | |||
XM_005273615.3 | XP_005273672.1 | |||
XM_006716390.3 | XP_006716453.1 | |||
XM_006716391.3 | XP_006716454.1 | |||
XM_017013748.1 | XP_016869237.1 | |||
XM_017013749.1 | XP_016869238.1 | |||
XM_017013750.1 | XP_016869239.1 | |||
XM_017013751.1 | XP_016869240.1 | |||
XM_017013752.1 | XP_016869241.1 | |||
AK291202.1 | ||||
AK295952.1 | ||||
AK314774.1 | ||||
BC028600.2 | AAH28600.1 | |||
L20852.1 | AAA18018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3617010 | Chr.8:42419628 - 42426318 on Build GRCh38 | Loss | SLC20A2 |
nsv396084 | Chr.8:42414652 - 42424106 on Build GRCh38 | Deletion | SLC20A2 |
nsv831301 | Chr.8:42244080 - 42473666 on Build GRCh38 | Loss | DKK4 VDAC3 LOC101929897 SLC20A2 IKBKB LOC105379395 POLB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |