Assay Details
Target Gene Details
Entrez Gene ID: | 55326 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 5 |
Gene Aliases: |
1AGPAT5, LPAATE |
Location: |
Chr.8:6708357-6761503 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGPAT5 | NM_018361.3 | NP_060831.2 | ||
XM_011534751.2 | XP_011533053.1 | |||
AF375789.1 | AAK54809.1 | |||
AK002072.1 | BAA92069.1 | |||
AK310545.1 | ||||
AL136587.1 | CAB66522.1 | |||
BC023550.2 | AAH23550.1 | |||
BC068519.1 | AAH68519.1 | |||
BC080537.1 | AAH80537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1021684 | Chr.8:6492570 - 6951933 on Build GRCh38 | Loss | MIR4659A ANGPT2 XKR5 MIR4659B AGPAT5 MIR8055 DEFA8P DEFB1 MCPH1-AS1 GS1-24F4.2 DEFA6 DEFA4 MCPH1 |
nsv610004 | Chr.8:6544845 - 6915391 on Build GRCh38 | Gain | MIR4659A MIR4659B AGPAT5 MIR8055 DEFB1 MCPH1-AS1 ANGPT2 XKR5 GS1-24F4.2 MCPH1 |
nsv1025976 | Chr.8:6615427 - 6829450 on Build GRCh38 | Loss | MIR4659A MIR4659B AGPAT5 MIR8055 MCPH1-AS1 XKR5 MCPH1 |
esv2422383 | Chr.8:6504868 - 6924071 on Build GRCh38 | Duplication | MIR4659A MIR4659B AGPAT5 MIR8055 DEFB1 MCPH1-AS1 ANGPT2 XKR5 GS1-24F4.2 MCPH1 |
nsv831220 | Chr.8:6670324 - 6870969 on Build GRCh38 | Gain | MIR4659A MIR4659B AGPAT5 DEFB1 MCPH1-AS1 XKR5 GS1-24F4.2 |
nsv610006 | Chr.8:6710680 - 7063882 on Build GRCh38 | Gain | MIR4659A DEFA7P DEFT1P DEFA5 DEFA11P DEFA9P XKR5 DEFA1 DEFA10P DEFA1B MIR4659B AGPAT5 DEFT1P2 DEFA8P DEFA3 DEFB1 GS1-24F4.2 DEFA6 DEFA4 |
nsv1031577 | Chr.8:6712833 - 8008628 on Build GRCh38 | Gain | DEFB108P1 USP17L3 DEFA11P DEFA9P USP17L4 USP17L1 ZNF705G DEFA6 SPAG11A LOC101929400 DEFB104A DEFB103B USP17L8 DEFA1B DEFB4A DEFA3 DEFB105B GS1-24F4.2 PRR23D1 OR7E154P FAM90A7P DEFB107A DEFA1 PRR23D2 DEFB106B DEFT1P2 DEFA8P DEFB103A OR7E125P LINC00965 DEFB104B FAM66E MIR4659A ZNF705B DEFB109P1B SPAG11B LOC107986910 LOC101928095 DEFA7P DEFB105A DEFB4B DEFT1P DEFA5 XKR5 DEFB106A DEFB107B DEFA10P MIR4659B AGPAT5 DEFB1 FAM66B FAM90A10P DEFA4 |
dgv6876n100 | Chr.8:6687977 - 6812428 on Build GRCh38 | Loss | MIR4659A MIR4659B AGPAT5 MCPH1-AS1 XKR5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |