Assay Details
Target Gene Details
Entrez Gene ID: | 286183 |
Gene Name: | Na+/K+ transporting ATPase interacting 3 |
Gene Aliases: |
FAM77D |
Location: |
Chr.8:62248942-62999652 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NKAIN3 | NM_001304533.1 | NP_001291462.1 | ||
NR_130764.1 | ||||
XM_011517511.2 | XP_011515813.1 | |||
XM_011517512.2 | XP_011515814.1 | |||
XM_017013359.1 | XP_016868848.1 | |||
XM_017013360.1 | XP_016868849.1 | |||
AK096949.1 | BAC04909.1 | |||
AK299689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1019143 | Chr.8:62229643 - 62478261 on Build GRCh38 | Loss | NKAIN3 |
esv3891406 | Chr.8:61776965 - 62523810 on Build GRCh38 | Gain | LOC101929628 NKAIN3 |
nsv1016883 | Chr.8:62198182 - 62258795 on Build GRCh38 | Gain | NKAIN3 |
esv2759621 | Chr.8:62243564 - 62347623 on Build GRCh38 | Loss | NKAIN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |