Assay Details
Target Gene Details
Entrez Gene ID: | 26 |
Gene Name: | amine oxidase, copper containing 1 |
Gene Aliases: |
ABP, ABP1, DAO, DAO1, KAO |
Location: |
Chr.7:150824875-150861291 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AOC1 | XM_017011944.1 | XP_016867433.1 | ||
XM_017011945.1 | XP_016867434.1 | |||
BX648159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1030109 | Chr.7:150615828 - 151003922 on Build GRCh38 | Gain | TMEM176B KCNH2 AOC1 GIMAP1 GIMAP2 GIMAP1-GIMAP5 GIMAP5 LOC105375566 NOS3 TMEM176A GIMAP6 |
esv2759574 | Chr.7:150841733 - 150966609 on Build GRCh38 | Loss | KCNH2 AOC1 |
nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain | AOC1 GIMAP1 GIMAP2 GIMAP1-GIMAP5 GIMAP5 GIMAP4 LOC105375566 NOS3 GIMAP7 REPIN1 ZBED6CL RARRES2 LRRC61 TMEM176B KCNH2 GIMAP8 LOC728743 LINC00996 ZNF775 LOC107986860 TMEM176A GIMAP6 ACTR3C |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |