Assay Details
Target Gene Details
Entrez Gene ID: | 2739 |
Gene Name: | glyoxalase I |
Gene Aliases: |
GLOD1, GLYI, HEL-S-74 |
Location: |
Chr.6:38675925-38703176 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLO1 | NM_006708.2 | NP_006699.2 | ||
AB209801.1 | BAD93038.1 | |||
AK129573.1 | ||||
AK130312.1 | ||||
AK293345.1 | ||||
AK312662.1 | ||||
AL833442.1 | ||||
BC001741.1 | AAH01741.1 | |||
BC011365.1 | AAH11365.1 | |||
BC015934.1 | AAH15934.1 | |||
BT019987.1 | AAV38790.1 | |||
BT019988.1 | AAV38791.1 | |||
D13315.1 | BAA02572.1 | |||
DA342581.1 | ||||
FJ224325.1 | ||||
KJ534846.1 | ||||
L07837.1 | AAA52565.1 | |||
S83285.1 | AAB49495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830642 | Chr.6:38677280 - 38832881 on Build GRCh38 | Loss | GLO1 DNAH8 |
nsv428144 | Chr.6:38621193 - 38743309 on Build GRCh38 | Gain | GLO1 BTBD9 DNAH8 |
nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain | PIM1 CPNE5 GLO1 LOC100131047 PI16 MIR3925 MIR4462 TBC1D22B LOC107986531 TMEM217 LOC100505530 SRSF3 GLP1R C6orf89 FGD2 CCDC167 MDGA1 BTBD9 PPIL1 MTCH1 RAB44 PANDAR ZFAND3 RNF8 DNAH8 CMTR1 CDKN1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |