Assay Details
Target Gene Details
Entrez Gene ID: | 285033 |
Gene Name: | STARD7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:96208416-96242623 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7-AS1 | NR_046322.1 | 4 | 2274 | |
AI218644.1 | 1 | 211 | ||
AK057419.1 | 4 | 1746 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | CIAO1 STARD7 SNRNP200 ITPRIPL1 LOC105373495 NCAPH TMEM127 STARD7-AS1 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | LOC100506123 LOC653924 GPAT2 ANKRD36 LOC107984110 TMEM127 STARD7-AS1 SEMA4C CNNM4 SNRNP200 DUSP2 ADRA2B ITPRIPL1 LOC105373495 MIR3127 LOC100506076 KANSL3 ANKRD39 CIAO1 ARID5A NCAPH LOC101927053 NEURL3 ASTL ANKRD36C LMAN2L STARD7 FAHD2B FER1L5 CNNM3 FAHD2CP ANKRD23 LOC105373496 FAM178B ANKRD36B |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | LOC100506123 LOC653924 GPAT2 ANKRD36 LOC107984110 TMEM127 STARD7-AS1 SEMA4C CNNM4 SNRNP200 DUSP2 ADRA2B ITPRIPL1 LOC105373495 MIR3127 LOC100506076 KANSL3 ANKRD39 CIAO1 ARID5A NCAPH LOC101927053 NEURL3 ASTL ANKRD36C LMAN2L STARD7 FAHD2B FER1L5 CNNM3 FAHD2CP ANKRD23 LOC105373496 FAM178B ANKRD36B |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | LOC100506123 LOC653924 GPAT2 ANKRD36 LOC107984110 TMEM127 STARD7-AS1 SEMA4C CNNM4 SNRNP200 DUSP2 ADRA2B ITPRIPL1 LOC105373495 MIR3127 LOC100506076 KANSL3 ANKRD39 CIAO1 ARID5A NCAPH LOC101927053 NEURL3 ASTL ANKRD36C LMAN2L STARD7 FAHD2B FER1L5 CNNM3 FAHD2CP ANKRD23 LOC105373496 FAM178B ANKRD36B |