Assay Details
Target Gene Details
Entrez Gene ID: | 30818 |
Gene Name: | potassium voltage-gated channel interacting protein 3 |
Gene Aliases: |
CSEN, DREAM, KCHIP3 |
Location: |
Chr.2:95297324-95386077 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNIP3 | NM_013434.4 | NP_038462.1 | ||
AF120102.1 | AAD20350.1 | |||
AF199599.1 | AAF33684.1 | |||
AF367022.1 | AAK53711.1 | |||
AJ131730.2 | CAB56835.1 | |||
AK315437.1 | ||||
BC012850.1 | AAH12850.1 | |||
BI755669.1 | ||||
BT020075.1 | AAV38878.1 | |||
DQ148485.1 | AAZ77802.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528011 | Chr.2:95265470 - 95391263 on Build GRCh38 | Loss | PROM2 KCNIP3 |
nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication | PROM2 ZNF2 MAL MRPS5 KCNIP3 FAHD2A ZNF514 |
esv2759076 | Chr.2:95304390 - 95486811 on Build GRCh38 | Loss | KCNIP3 FAHD2A TRIM43B |
nsv998014 | Chr.2:95247530 - 95630882 on Build GRCh38 | Gain | PROM2 TRIM43 KCNIP3 FAHD2A LOC100419920 TRIM43B TRIM64FP TRIM51JP |
nsv953478 | Chr.2:95266753 - 95326752 on Build GRCh38 | Deletion | PROM2 KCNIP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |