Assay Details
Target Gene Details
Entrez Gene ID: | 23433 |
Gene Name: | ras homolog family member Q |
Gene Aliases: |
ARHQ, HEL-S-42, RASL7A, TC10, TC10A |
Location: |
Chr.2:46542409-46584688 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RHOQ | NM_012249.3 | NP_036381.2 | ||
XM_005264229.2 | XP_005264286.1 | |||
XM_011532726.2 | XP_011531028.1 | |||
XM_011532728.2 | XP_011531030.1 | |||
XM_017003716.1 | XP_016859205.1 | |||
AF498976.1 | AAM21123.1 | |||
AK095329.1 | ||||
BC056154.1 | ||||
BC065291.1 | ||||
BC070485.1 | AAH70485.2 | |||
BC093805.1 | AAH93805.2 | |||
BC101806.1 | ||||
EU794657.1 | ||||
M31470.1 | AAA36547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3590613 | Chr.2:46548390 - 46562976 on Build GRCh38 | Gain | RHOQ |
esv2759046 | Chr.2:46305673 - 46647012 on Build GRCh38 | Loss | RHOQ PIGF TMEM247 LOC101805491 ATP6V1E2 EPAS1 LOC100506142 CRIPT |
nsv9713 | Chr.2:46548217 - 46563427 on Build GRCh38 | Gain | RHOQ |
dgv149e55 | Chr.2:46378520 - 46591853 on Build GRCh38 | Loss | RHOQ PIGF TMEM247 LOC101805491 ATP6V1E2 EPAS1 LOC100506142 |
nsv1009735 | Chr.2:46497384 - 46686013 on Build GRCh38 | Loss | RHOQ PIGF ATP6V1E2 LOC100506142 CRIPT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |