Assay Details
Target Gene Details
Entrez Gene ID: | 285033 |
Gene Name: | STARD7 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:96208416-96242623 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7-AS1 | NR_046322.1 | |||
AK057419.1 | ||||
AK311432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv963665 | Chr.2:96216515 - 96239873 on Build GRCh38 | Duplication | STARD7-AS1 |
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | SNRNP200 STARD7-AS1 NCAPH LOC105373495 ITPRIPL1 STARD7 TMEM127 CIAO1 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | TMEM127 LOC100506123 LOC100506076 ANKRD23 MIR3127 STARD7-AS1 NCAPH STARD7 LOC107984110 FER1L5 ASTL LOC653924 ANKRD36C SNRNP200 LOC101927053 ANKRD36B SEMA4C FAHD2B ARID5A FAM178B LMAN2L LOC105373496 CNNM4 GPAT2 KANSL3 NEURL3 DUSP2 ANKRD36 ADRA2B LOC105373495 ITPRIPL1 ANKRD39 CIAO1 FAHD2CP CNNM3 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | TMEM127 LOC100506123 LOC100506076 ANKRD23 MIR3127 STARD7-AS1 NCAPH STARD7 LOC107984110 FER1L5 ASTL LOC653924 ANKRD36C SNRNP200 LOC101927053 ANKRD36B SEMA4C FAHD2B ARID5A FAM178B LMAN2L LOC105373496 CNNM4 GPAT2 KANSL3 NEURL3 DUSP2 ANKRD36 ADRA2B LOC105373495 ITPRIPL1 ANKRD39 CIAO1 FAHD2CP CNNM3 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | TMEM127 LOC100506123 LOC100506076 ANKRD23 MIR3127 STARD7-AS1 NCAPH STARD7 LOC107984110 FER1L5 ASTL LOC653924 ANKRD36C SNRNP200 LOC101927053 ANKRD36B SEMA4C FAHD2B ARID5A FAM178B LMAN2L LOC105373496 CNNM4 GPAT2 KANSL3 NEURL3 DUSP2 ANKRD36 ADRA2B LOC105373495 ITPRIPL1 ANKRD39 CIAO1 FAHD2CP CNNM3 |