Assay Details
Target Gene Details
Entrez Gene ID: | 55733 |
Gene Name: | hedgehog acyltransferase |
Gene Aliases: |
MART2, SKI1, Skn |
Location: |
Chr.1:210327873-210676298 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv8769 | Chr.1:210230164 - 210391244 on Build GRCh38 | Gain | HHAT SERTAD4 SERTAD4-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |