Assay Details
Target Gene Details
Entrez Gene ID: | 375056 |
Gene Name: | MIA family member 3, ER export factor |
Gene Aliases: |
ARNT, D320, TANGO, TANGO1, UNQ6077 |
Location: |
Chr.1:222617790-222668012 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MIA3 | NM_001300867.1 | 23 | 2949 | NP_001287796.1 |
NM_001324062.1 | 29 | 6223 | NP_001310991.1 | |
NM_001324063.1 | 26 | 5974 | NP_001310992.1 | |
NM_001324064.1 | 29 | 6196 | NP_001310993.1 | |
NM_001324065.1 | 24 | 3021 | NP_001310994.1 | |
NM_198551.3 | 28 | 6151 | NP_940953.2 | |
XM_006711304.3 | 27 | 6358 | XP_006711367.2 | |
XM_017001243.1 | 29 | 6048 | XP_016856732.1 | |
AK026489.1 | 17 | 2210 | ||
AY359091.1 | 5 | 1913 | AAQ89449.1 | |
D87742.1 | 25 | 4000 | BAA13448.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74822125] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |