Assay Details
Target Gene Details
Entrez Gene ID: | 81539 |
Gene Name: | solute carrier family 38 member 1 |
Gene Aliases: |
ATA1, NAT2, SAT1, SNAT1 |
Location: |
Chr.12:46183055-46269425 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC38A1 | NM_001077484.1 | 17 | 2598 | NP_001070952.1 |
NM_001278387.1 | 18 | 3060 | NP_001265316.1 | |
NM_001278388.1 | 16 | 2822 | NP_001265317.1 | |
NM_001278389.1 | 17 | 2087 | NP_001265318.1 | |
NM_030674.3 | 17 | 2937 | NP_109599.3 | |
AF247166.1 | 17 | 2282 | AAG44546.1 | |
AF271070.1 | 17 | 2181 | AAG39354.1 | |
AK027825.1 | 17 | 2087 | BAB55394.1 | |
AK074758.1 | 10 | 1354 | BAC11186.1 | |
AK074797.1 | 14 | 1656 | BAC11215.1 | |
AK074851.1 | 15 | 1799 | ||
AK074949.1 | 15 | 1845 | BAC11310.1 | |
AK225639.1 | 17 | 2175 | ||
AK225778.1 | 17 | 2197 | ||
BC010620.1 | 17 | 2144 | AAH10620.1 | |
BX647547.1 | 18 | 2632 | ||
BX648495.1 | 17 | 2525 | ||
BX648676.1 | 16 | 2392 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv826360 | Chr.12:46121591 - 46204460 on Build GRCh38 | Loss | SLC38A1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |