Assay Details
Target Gene Details
Entrez Gene ID: | 339229 |
Gene Name: | oxidoreductase like domain containing 1 |
Gene Aliases: |
C17orf90 |
Location: |
Chr.17:81665036-81666637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OXLD1 | NM_001039842.2 | 2 | 190 | NP_001034931.1 |
NM_001304994.1 | 2 | 544 | NP_001291923.1 | |
NM_001304995.1 | 2 | 431 | NP_001291924.1 | |
NM_001304999.1 | 1 | 1123 | NP_001291928.1 | |
NR_130936.1 | 2 | 553 | ||
NR_130937.1 | 2 | 558 | ||
NR_130938.1 | 2 | 562 | ||
NR_130939.1 | 2 | 219 | ||
AY007126.1 | 1 | 1121 | ||
BC090923.1 | 2 | 141 | AAH90923.1 | |
BE386163.1 | 2 | 505 | ||
BU194770.1 | 2 | 389 | ||
CB048026.1 | 2 | 227 | ||
DT220680.1 | 2 | 499 | ||
HY012937.1 | ||||
HY056467.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC105371921 SLC25A10 MIR4740 LOC105371925 HGS FSCN2 MRPL12 LOC400627 NPLOC4 CHMP6 BAHCC1 RPTOR OXLD1 TSPAN10 TMEM105 MIR3186 FAAP100 MIR657 ACTG1 MIR6786 AATK-AS1 LOC100129503 CCDC137 TEPSIN C17orf89 AATK BAIAP2 MIR338 MIR3065 LOC107985021 LOC101928855 LINC00482 CEP131 LOC107985022 SLC38A10 ARL16 LOC100130370 BAIAP2-AS1 MIR1250 PDE6G |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR4525 FOXK2 MAFG-AS1 SLC25A10 LOC105371925 PYCR1 ZNF750 OGFOD3 SLC16A3 HGS FSCN2 MIR6787 NPLOC4 BAHCC1 FAAP100 ARHGDIA ACTG1 AATK-AS1 C17orf62 TEX19 UTS2R LOC101929552 DCXR ASPSCR1 LOC105376790 TBCD B3GNTL1 LOC101929511 MYADML2 LINC00482 DUS1L CEP131 ANAPC11 ARL16 LOC100130370 LRRC45 MCRIP1 HEXDC CSNK1D LOC105371944 PDE6G WDR45B CCDC57 CD7 MIR4740 P4HB GCGR ALYREF RFNG RAB40B MRPL12 GPS1 OXLD1 TSPAN10 TMEM105 MIR3186 NPB MIR6786 PPP1R27 FN3KRP RAC3 CCDC137 SIRT7 NARF TEPSIN STRA13 C17orf89 NOTUM FN3K AATK SECTM1 SNORD134 LOC105376791 MAFG SLC38A10 FASN PCYT2 |
nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss | MAFG-AS1 SLC25A10 PYCR1 P4HB GCGR HGS FSCN2 ALYREF RFNG MRPL12 NPLOC4 GPS1 OXLD1 TSPAN10 FAAP100 ARHGDIA NPB MIR6786 PPP1R27 RAC3 CCDC137 SIRT7 DCXR ASPSCR1 STRA13 NOTUM MYADML2 DUS1L ANAPC11 MAFG ARL16 LRRC45 MCRIP1 PDE6G PCYT2 |
nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion | SLC25A10 P4HB GCGR HGS MRPL12 OXLD1 TSPAN10 MIR6786 PPP1R27 ARL16 CCDC137 MCRIP1 PDE6G |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |