Assay Details
Target Gene Details
Entrez Gene ID: | 23543 |
Gene Name: | RNA binding protein, fox-1 homolog 2 |
Gene Aliases: |
FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh |
Location: |
Chr.22:35738736-36028892 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3612 | Chr.22:35713114 - 35761709 on Build GRCh38 | Insertion | RBFOX2 APOL5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |