Assay Details
Target Gene Details
Entrez Gene ID: | 51126 |
Gene Name: | N(alpha)-acetyltransferase 20, NatB catalytic subunit |
Gene Aliases: |
NAT3, NAT3P, NAT5, NAT5P, dJ1002M8.1 |
Location: |
Chr.20:20017290-20033629 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA20 | NM_016100.4 | 6 | 896 | NP_057184.1 |
NM_181527.3 | 6 | 983 | NP_852668.1 | |
NM_181528.3 | 5 | 750 | NP_852669.1 | |
AF085355.1 | 6 | 859 | AAD40190.1 | |
AL136641.1 | 6 | 828 | CAB66576.1 | |
BC005181.1 | 6 | 824 | AAH05181.1 | |
BC008446.1 | 6 | 845 | AAH08446.1 | |
BG548527.1 | ||||
BU680824.1 | 1 | 207 | ||
BX338359.1 | 1 | 130 | ||
CA952537.1 | 1 | 468 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |