Assay Details
Target Gene Details
Entrez Gene ID: | 10524 |
Gene Name: | lysine acetyltransferase 5 |
Gene Aliases: |
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2 |
Location: |
Chr.11:65711996-65719606 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAT5 | NM_001206833.1 | NP_001193762.1 | ||
NM_006388.3 | NP_006379.2 | |||
NM_182709.2 | NP_874368.1 | |||
NM_182710.2 | NP_874369.1 | |||
XM_006718421.2 | XP_006718484.1 | |||
AB209813.1 | BAD93050.1 | |||
AK225100.1 | ||||
AK226002.1 | ||||
AK304664.1 | ||||
AK313297.1 | ||||
BC000166.3 | AAH00166.3 | |||
BC064912.1 | AAH64912.1 | |||
BC093032.1 | AAH93032.1 | |||
BC117167.1 | ||||
BC143296.1 | ||||
BG720537.1 | ||||
BI463394.1 | ||||
DA043042.1 | ||||
DB003934.1 | ||||
DB103287.1 | ||||
U40989.1 | AAB02683.1 | |||
U67734.1 | AAD00163.1 | |||
U74667.1 | AAB18236.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |