Assay Details
Target Gene Details
Entrez Gene ID: | 50604 |
Gene Name: | interleukin 20 |
Gene Aliases: |
IL-20, IL10D, ZCYTO10 |
Location: |
Chr.1:206865247-206869223 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL20 | NM_018724.3 | NP_061194.2 | ||
XM_005273152.3 | XP_005273209.1 | |||
AF224266.1 | AAF36679.1 | |||
AY358320.1 | AAQ88686.1 | |||
AY876922.1 | AAW78348.1 | |||
BC069311.1 | AAH69311.1 | |||
BC069364.1 | AAH69364.1 | |||
BC069398.1 | AAH69398.1 | |||
BC069425.1 | AAH69425.1 | |||
BC069449.1 | AAH69449.1 | |||
BC069487.1 | AAH69487.1 | |||
BC069523.1 | AAH69523.1 | |||
BC069559.1 | AAH69559.1 | |||
BC074948.2 | AAH74948.1 | |||
BC074949.2 | AAH74949.1 | |||
BC117386.1 | ||||
BC117390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549070 | Chr.1:206857640 - 206867314 on Build GRCh38 | Loss | IL20 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |