Assay Details
Target Gene Details
Entrez Gene ID: | 55662 |
Gene Name: | hypoxia inducible factor 1 alpha subunit inhibitor |
Gene Aliases: |
FIH1 |
Location: |
Chr.10:100535884-100553924 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1AN | NM_017902.2 | NP_060372.2 | ||
XM_011539940.2 | XP_011538242.1 | |||
XM_011539941.1 | XP_011538243.1 | |||
AF395830.1 | AAL27308.1 | |||
AK000622.1 | BAA91291.1 | |||
AK025680.1 | BAB15213.1 | |||
AK295353.1 | ||||
AL359615.1 | CAB94885.1 | |||
BC007719.2 | AAH07719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831963 | Chr.10:100458231 - 100678922 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv831962 | Chr.10:100406551 - 100593251 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv2759779 | Chr.10:100392093 - 100805979 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)