Assay Details
Target Gene Details
Entrez Gene ID: | 124857 |
Gene Name: | WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 |
Gene Aliases: |
GASP-1, WFDC20B, WFIKKNRP, hGASP-1 |
Location: |
Chr.17:50834650-50842353 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFIKKN2 | NM_175575.5 | 1 | 633 | NP_783165.1 |
XM_017024172.1 | XP_016879661.1 | |||
AK127743.1 | 1 | 293 | ||
AK298172.1 | ||||
AK312821.1 | 1 | 125 | ||
AL537450.3 | 1 | 398 | ||
AY358142.1 | 1 | 467 | AAQ88509.1 | |
BC140842.1 | ||||
BC140844.1 | ||||
BQ954191.1 | 2 | 525 | ||
DB235333.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |