Assay Details
Target Gene Details
Entrez Gene ID: | 101929140 |
Gene Name: | uncharacterized LOC101929140 |
Gene Aliases: |
- |
Location: |
Chr.13:41132928-41236686 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929140 | NR_120423.1 | |||
AK056182.1 | ||||
DA634395.1 |
Target Gene Details
Entrez Gene ID: | 9617 |
Gene Name: | mitochondrial translational release factor 1 |
Gene Aliases: |
MRF1, MTTRF1, RF1 |
Location: |
Chr.13:41216369-41311794 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832587 | Chr.13:41030795 - 41224672 on Build GRCh38 | Gain | KBTBD7 MIR3168 MTRF1 KBTBD6 LOC101929140 WBP4 ELF1 |
nsv1014 | Chr.13:41196977 - 41229583 on Build GRCh38 | Insertion | MTRF1 LOC101929140 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |