Assay Details
Target Gene Details
Entrez Gene ID: | 54882 |
Gene Name: | ankyrin repeat and KH domain containing 1 |
Gene Aliases: |
MASK, MASK1, PP2500, VBARP |
Location: |
Chr.5:140401814-140539856 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANKHD1 | NM_001197030.1 | 10 | 1963 | NP_001183959.1 |
NM_017747.2 | NP_060217.1 | |||
NM_017978.2 | 11 | 2040 | NP_060448.1 | |
NM_024668.3 | 11 | 2073 | NP_078944.2 | |
AF258557.1 | 11 | 2033 | AAG23760.1 | |
AF521882.1 | AAO14943.1 | |||
AK000295.1 | BAA91063.1 | |||
AK022041.1 | 10 | 1609 | BAB13958.1 | |
AK074173.1 | BAB84999.1 | |||
AK098574.1 | ||||
AY070436.1 | AAL65263.1 | |||
BC004457.1 | 11 | 2018 | AAH04457.2 | |
BC009420.1 | 11 | 1933 | AAH09420.1 | |
BC009909.2 | 11 | 2018 | AAH09909.1 | |
BC117677.2 | 11 | 1999 | ||
BC117678.2 | 10 | 1889 | ||
BC127127.1 | 11 | 1999 | ||
BC150486.2 | 11 | 2003 |
Target Gene Details
Entrez Gene ID: | 404734 |
Gene Name: | ANKHD1-EIF4EBP3 readthrough |
Gene Aliases: |
MASK-BP3, MASK-BP3ARF |
Location: |
Chr.5:140401814-140549578 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANKHD1-EIF4EBP3 | NM_020690.5 | NP_065741.3 | ||
AF521883.1 | AAO14944.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |