Assay Details
Target Gene Details
Entrez Gene ID: | 80781 |
Gene Name: | collagen type XVIII alpha 1 chain |
Gene Aliases: |
KNO, KNO1, KS |
Location: |
Chr.21:45405137-45513720 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COL18A1 | NM_130445.3 | NP_569712.2 | ||
AF018082.1 | AAC39659.1 | |||
AK130835.1 | ||||
BC063833.1 | AAH63833.1 | |||
CN389577.1 |
Target Gene Details
Entrez Gene ID: | 378832 |
Gene Name: | COL18A1 antisense RNA 1 |
Gene Aliases: |
C21orf123, NCRNA00175, PRED80 |
Location: |
Chr.21:45419716-45425070 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COL18A1-AS1 | NR_027498.1 | 3 | 2131 | |
NR_028082.1 | 3 | 3215 | ||
AK096071.1 | 3 | 2131 | BAC04694.1 | |
BC111539.2 | 3 | 3215 | AAI11540.1 | |
BC111540.2 | 3 | 2101 | AAI11541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516407 | Chr.21:45410958 - 45620742 on Build GRCh38 | Loss | COL18A1-AS1 SLC19A1 MIR6815 LOC105372840 COL18A1 |
esv2762114 | Chr.21:45260287 - 45647830 on Build GRCh38 | Gain | LINC00205 POFUT2 LOC642852 LINC00316 COL18A1-AS1 SLC19A1 MIR6815 COL18A1-AS2 LOC105372840 LOC107985476 COL18A1 |
nsv953653 | Chr.21:45409686 - 45560386 on Build GRCh38 | Deletion | COL18A1-AS1 SLC19A1 MIR6815 COL18A1-AS2 COL18A1 |
nsv1057607 | Chr.21:45333418 - 45463244 on Build GRCh38 | Gain | LINC00316 COL18A1-AS1 COL18A1-AS2 COL18A1 |
dgv4436n100 | Chr.21:45402338 - 45556768 on Build GRCh38 | Gain | COL18A1-AS1 SLC19A1 MIR6815 COL18A1-AS2 COL18A1 |
dgv7899n54 | Chr.21:45160455 - 45419989 on Build GRCh38 | Gain | LINC00205 POFUT2 LOC642852 LINC00334 LINC00316 COL18A1-AS1 COL18A1-AS2 ADARB1 LOC107985476 COL18A1 |
nsv470909 | Chr.21:45300095 - 45428674 on Build GRCh38 | Gain | LINC00316 COL18A1-AS1 COL18A1-AS2 COL18A1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |