Assay Details
Target Gene Details
Entrez Gene ID: | 10524 |
Gene Name: | lysine acetyltransferase 5 |
Gene Aliases: |
ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, ZC2HC5, cPLA2 |
Location: |
Chr.11:65711996-65719606 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KAT5 | NM_001206833.1 | 12 | 1997 | NP_001193762.1 |
NM_006388.3 | 14 | 2054 | NP_006379.2 | |
NM_182709.2 | 13 | 1898 | NP_874368.1 | |
NM_182710.2 | 13 | 2153 | NP_874369.1 | |
XM_006718421.2 | 14 | 2087 | XP_006718484.1 | |
AB209813.1 | 11 | 1700 | BAD93050.1 | |
AK225100.1 | 14 | 1739 | ||
AK226002.1 | 14 | 1739 | ||
AK304664.1 | ||||
BC000166.3 | 12 | 1620 | AAH00166.3 | |
BC064912.1 | 14 | 1794 | AAH64912.1 | |
BC093032.1 | 13 | 1665 | AAH93032.1 | |
BU737532.1 | 1 | 195 | ||
U40989.1 | 13 | 1759 | AAB02683.1 | |
U67734.1 | 13 | 1649 | AAD00163.1 | |
U74667.1 | 14 | 2029 | AAB18236.1 |
Target Gene Details
Entrez Gene ID: | 84153 |
Gene Name: | ribonuclease H2 subunit C |
Gene Aliases: |
AGS3, AYP1 |
Location: |
Chr.11:65717673-65720938 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNASEH2C | NM_032193.3 | 4 | 1031 | NP_115569.2 |
AK297660.1 | 4 | 801 | ||
BM921525.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |