Assay Details
Target Gene Details
Entrez Gene ID: | 51296 |
Gene Name: | solute carrier family 15 member 3 |
Gene Aliases: |
OCTP, PHT2, PTR3 |
Location: |
Chr.11:60937083-60952081 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC15A3 | NM_016582.2 | 8 | 2048 | NP_057666.1 |
NR_027391.1 | 7 | 1879 | ||
XM_011545095.2 | 9 | 2394 | XP_011543397.1 | |
AB020598.1 | 8 | 2048 | BAA93432.1 | |
AF135600.1 | 8 | 1941 | AAQ13565.1 | |
AK127216.1 | 3 | 4503 | ||
BC025710.1 | 7 | 1739 | ||
BC037974.1 | 8 | 1908 | AAH37974.1 | |
BU731752.1 | 1 | 87 |
Target Gene Details
Entrez Gene ID: | 54972 |
Gene Name: | transmembrane protein 132A |
Gene Aliases: |
GBP, HSPA5BP1 |
Location: |
Chr.11:60924441-60937159 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM132A | NM_017870.3 | NP_060340.2 | ||
NM_178031.2 | NP_821174.1 | |||
XM_017017951.1 | XP_016873440.1 | |||
XM_017017952.1 | XP_016873441.1 | |||
AK000546.1 | BAA91245.1 | |||
AK023577.1 | BAB14613.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |