Assay Details
Target Gene Details
Entrez Gene ID: | 8996 |
Gene Name: | nucleolar protein 3 |
Gene Aliases: |
ARC, FCM, MYP, NOP, NOP30 |
Location: |
Chr.16:67170497-67175737 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOL3 | NM_001185057.2 | 2 | 352 | NP_001171986.1 |
NM_001276307.1 | 4 | 376 | NP_001263236.1 | |
NM_001276309.1 | 2 | 236 | NP_001263238.1 | |
NM_001276311.1 | NP_001263240.1 | |||
NM_001276312.1 | 4 | 727 | NP_001263241.1 | |
NM_001276319.1 | 4 | 727 | NP_001263248.1 | |
NM_003946.6 | 2 | 352 | NP_003937.1 | |
XM_005256217.2 | 2 | 989 | XP_005256274.1 | |
XM_005256219.3 | XP_005256276.1 | |||
XM_011523424.2 | 3 | 832 | XP_011521726.1 | |
XM_017023843.1 | 2 | 989 | XP_016879332.1 | |
AF043244.1 | 2 | 269 | AAC34993.1 | |
AF064599.1 | 2 | 203 | AAC18593.1 | |
AF064600.1 | 2 | 227 | AAC18594.1 | |
AK294145.1 | 4 | 692 | ||
AK314899.1 | 2 | 257 | ||
BC012798.2 | 2 | 238 | AAH12798.1 | |
BP215200.1 | 2 | 193 | ||
BQ685512.1 | 2 | 352 | ||
BT020158.1 | 1 | 150 | AAV38960.1 | |
CB959673.1 | 4 | 403 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | DPEP3 PARD6A HSF4 LOC107984813 LRRC36 DUS2 NUTF2 ACD CENPT HSD11B2 NRN1L DPEP2 LCAT LOC105369155 LOC100505942 TPPP3 RANBP10 KCTD19 ATP6V0D1 PSKH1 DDX28 MIR328 C16orf86 ENKD1 CARMIL2 GFOD2 FAM65A PLEKHG4 CTCF CTRL LOC100131303 THAP11 NOL3 TRADD ELMO3 ZDHHC1 PSMB10 SLC9A5 AGRP LRRC29 EXOC3L1 SLC12A4 E2F4 TSNAXIP1 EDC4 FHOD1 KIAA0895L TMEM208 FBXL8 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | SLC9A5 HSF4 LOC105369155 LRRC29 EXOC3L1 E2F4 MIR328 C16orf70 B3GNT9 PLEKHG4 FHOD1 KIAA0895L TMEM208 FBXL8 NOL3 TRADD ELMO3 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | HSF4 LOC105369155 LRRC29 EXOC3L1 E2F4 MIR328 C16orf70 B3GNT9 FHOD1 KIAA0895L TMEM208 FBXL8 NOL3 TRADD ELMO3 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | MIR328 HSF4 KIAA0895L LOC105369155 LRRC29 EXOC3L1 FBXL8 NOL3 ELMO3 E2F4 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | SLC9A5 LOC105369155 LRRC29 EXOC3L1 E2F4 MIR328 KCTD19 PLEKHG4 FHOD1 KIAA0895L TMEM208 NOL3 ELMO3 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | DPEP3 PARD6A HSF4 LOC107984813 LRRC36 DUS2 NUTF2 C16orf70 ACD CENPT HSD11B2 NRN1L DPEP2 LCAT LOC105369155 LOC100505942 TPPP3 RANBP10 KCTD19 ATP6V0D1 PSKH1 DDX28 MIR328 C16orf86 ENKD1 B3GNT9 CARMIL2 GFOD2 FAM65A PLEKHG4 CTCF CTRL LOC100131303 THAP11 NOL3 TRADD ELMO3 ZDHHC1 PSMB10 SLC9A5 AGRP LRRC29 EXOC3L1 SLC12A4 E2F4 TSNAXIP1 EDC4 FHOD1 KIAA0895L TMEM208 FBXL8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |