Assay Details
Target Gene Details
Entrez Gene ID: | 1355 |
Gene Name: | COX15, cytochrome c oxidase assembly homolog |
Gene Aliases: |
CEMCOX2 |
Location: |
Chr.10:99694793-99732667 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX15 | NM_001320974.1 | NP_001307903.1 | ||
NM_001320975.1 | 8 | 1998 | NP_001307904.1 | |
NM_001320976.1 | 9 | 2070 | NP_001307905.1 | |
NM_004376.6 | NP_004367.2 | |||
NM_078470.5 | 9 | 2153 | NP_510870.1 | |
XM_006717634.3 | XP_006717697.1 | |||
XM_011539298.2 | XP_011537600.1 | |||
XM_017015715.1 | XP_016871204.1 | |||
AF026850.1 | 9 | 1587 | AAD08639.1 | |
AF044323.1 | AAD08646.1 | |||
AK291654.1 | ||||
BC002382.2 | 9 | 1552 | AAH02382.3 | |
BC013403.2 | 9 | 1552 | AAH13403.1 | |
BC078161.1 | 9 | 1564 | AAH78161.1 | |
BX537557.1 | 9 | 2153 | CAD97781.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |