Assay Details
Target Gene Details
Entrez Gene ID: | 57333 |
Gene Name: | reticulocalbin 3 |
Gene Aliases: |
RLP49 |
Location: |
Chr.19:49527579-49543638 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RCN3 | NM_020650.2 | 1 | 114 | NP_065701.2 |
XM_005259089.3 | 1 | 153 | XP_005259146.1 | |
XM_011527143.1 | 1 | 153 | XP_011525445.1 | |
XM_017027023.1 | 1 | 153 | XP_016882512.1 | |
AY195859.1 | 1 | 114 | AAO43054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-A1 DBP SYNGR4 FUT1 LIN7B DHDH SNAR-B1 LOC100287477 CCDC155 SNORD33 ALDH16A1 ADM5 HRC FUT2 LOC105447645 SNAR-A10 FCGRT PTOV1-AS2 RCN3 RUVBL2 BCAT2 FTL LOC101928295 SNAR-A4 GRWD1 PLEKHA4 CGB7 CGB5 CPT1C PRMT1 CGB1 FLJ26850 MIR4749 SNAR-A6 LOC105372430 SNAR-G2 SLC6A16 TSKS NTF4 SNRNP70 CARD8 PTOV1-AS1 CGB3 CA11 NUCB1 RPL18 MIR4750 PTH2 KDELR1 PLA2G4C-AS1 SNAR-A14 SPACA4 DKKL1 MIR6799 SNAR-C4 NTN5 SNAR-C1 SLC17A7 C19orf73 RRAS SNAR-G1 CGB2 SNAR-A2 MIR4751 SIGLEC11 RPS11 MED25 SNAR-A9 SNAR-A11 SNORD35A LHB CARD8-AS1 LMTK3 SNORD35B SNAR-C2 SNORD32A SCAF1 CCDC114 TBC1D17 SIGLEC16 MAMSTR SNORD34 SNAR-C5 VRK3 RPL13A PLA2G4C SNAR-A3 NOSIP C19orf68 RASIP1 TULP2 MIR6798 NUCB1-AS1 KCNA7 SULT2B1 PIH1D1 LIG1 KCNJ14 GYS1 PRR12 FUZ SNAR-C3 ELSPBP1 PTOV1 NUP62 CABP5 MIR5088 MIR4324 EMP3 PRRG2 IZUMO1 TRPM4 MIR6800 PPFIA3 CD37 SNAR-A7 CGB8 HSD17B14 PPP1R15A TEAD2 SPHK2 IL4I1 GFY ZNF473 AP2A1 LOC101059948 MIR150 SNAR-D SEC1P CYTH2 SNAR-B2 FAM83E SNAR-A8 GRIN2D ZNF114 TMEM143 FGF21 ATF5 BSPH1 SNAR-A12 SNAR-A5 SNAR-A13 BCL2L12 FLT3LG BAX PNKP IRF3 AKT1S1 |
nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss | RCN3 MIR150 SNORD35A NOSIP SNORD35B SNORD32A PRRG2 SNORD33 SNORD34 RPL13A FLT3LG RPS11 PRR12 FCGRT |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | RCN3 MIR150 SNORD35A NOSIP SNORD35B SNORD32A SNORD33 ALDH16A1 SNORD34 PIH1D1 RPL13A FLT3LG RPS11 FCGRT |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | SNORD35A MIR5088 SNORD35B SNORD32A PRRG2 SCAF1 MIR6800 SNORD33 SNORD34 TSKS ADM5 RPL13A FCGRT PTOV1-AS1 RCN3 AP2A1 MIR150 MIR6799 NOSIP RRAS BCL2L12 CPT1C RPS11 IRF3 PRR12 MED25 PRMT1 FUZ |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |