Assay Details
Target Gene Details
Entrez Gene ID: | 116461 |
Gene Name: | tRNA splicing endonuclease subunit 15 |
Gene Aliases: |
C1orf19, PCH2F, sen15 |
Location: |
Chr.1:184050207-184095846 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSEN15 | NM_001127394.3 | NP_001120866.1 | ||
NM_001300764.1 | NP_001287693.1 | |||
NM_001300766.1 | NP_001287695.1 | |||
NM_052965.3 | NP_443197.1 | |||
NR_023349.2 | ||||
NR_125335.1 | ||||
XM_006711148.1 | XP_006711211.1 | |||
XM_011509139.2 | XP_011507441.1 | |||
XM_017000228.1 | XP_016855717.1 | |||
XM_017000229.1 | XP_016855718.1 | |||
XM_017000230.1 | XP_016855719.1 | |||
AF288394.1 | AAG60614.1 | |||
AK098196.1 | ||||
AK296655.1 | ||||
AK307241.1 | ||||
BC022030.1 | AAH22030.1 | |||
BI821111.1 | ||||
BM887891.1 | ||||
BP326115.1 | ||||
BU195337.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |