Assay Details
Target Gene Details
Entrez Gene ID: | 26097 |
Gene Name: | chromatin target of PRMT1 |
Gene Aliases: |
C1orf77, FL-SRAG, FOP, SRAG, SRAG-3, SRAG-5, pp7704 |
Location: |
Chr.1:153633982-153646306 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHTOP | NM_001206612.1 | NP_001193541.1 | ||
NM_001244664.1 | NP_001231593.1 | |||
NM_001317077.1 | NP_001304006.1 | |||
NM_015607.3 | NP_056422.2 | |||
XM_017000993.1 | XP_016856482.1 | |||
AF261137.1 | AAG44673.1 | |||
AF318362.1 | AAL55869.1 | |||
AK095230.1 | ||||
AK308713.1 | ||||
AK309844.1 | ||||
BC120961.1 | ||||
BC120962.1 | ||||
BP375397.1 | ||||
DC387712.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |