Assay Details
Target Gene Details
Entrez Gene ID: | 5931 |
Gene Name: | RB binding protein 7, chromatin remodeling factor |
Gene Aliases: |
RbAp46 |
Location: |
Chr.X:16844650-16870411 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBBP7 | NM_001198719.1 | 6 | 1037 | NP_001185648.1 |
NM_002893.3 | 6 | 1049 | NP_002884.1 | |
AK091911.1 | 6 | 1034 | ||
AK291567.1 | 6 | 793 | ||
AK310309.1 | 3 | 338 | ||
BC114500.1 | 6 | 947 | ||
BC114501.1 | 6 | 947 | ||
BQ775345.1 | 4 | 679 | ||
CR541701.1 | 6 | 691 | CAG46502.1 | |
U35143.1 | 6 | 952 | AAC50231.1 | |
X72841.1 | 6 | 976 | CAA51360.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |