Assay Details
Target Gene Details
Entrez Gene ID: | 51247 |
Gene Name: | poly(A) binding protein interacting protein 2 |
Gene Aliases: |
PAIP-2, PAIP2A |
Location: |
Chr.5:139341832-139369720 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAIP2 | NM_001033112.2 | NP_001028284.1 | ||
NM_016480.4 | NP_057564.3 | |||
NR_109830.1 | ||||
XM_011543428.1 | XP_011541730.1 | |||
XM_017009522.1 | 1 | 792 | XP_016865011.1 | |
XM_017009523.1 | XP_016865012.1 | |||
AF151052.1 | AAF36138.1 | |||
AF317675.1 | AAK11562.1 | |||
AK024552.1 | ||||
AK314672.1 | ||||
AL136640.1 | CAB66575.1 | |||
BC001716.1 | AAH01716.1 | |||
BC039337.1 | 1 | 132 | AAH39337.1 | |
BC048106.1 | 1 | 201 | AAH48106.1 | |
BC062718.1 | AAH62718.1 | |||
BI552800.1 | ||||
BI560379.1 | ||||
DB448759.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |