Assay Details
Target Gene Details
Entrez Gene ID: | 112398 |
Gene Name: | egl-9 family hypoxia inducible factor 2 |
Gene Aliases: |
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1 |
Location: |
Chr.19:40799143-40808441 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EGLN2 | NM_053046.3 | 2 | 510 | NP_444274.1 |
NM_080732.3 | 2 | 420 | NP_542770.2 | |
AJ310544.1 | 1 | 363 | CAC42510.1 | |
AK074408.1 | 2 | 349 | ||
AK098182.1 | 1 | 1094 | ||
AL832506.1 | 2 | 376 | ||
AY040565.1 | 1 | 354 | AAK82943.1 | |
BC001723.1 | 2 | 323 | AAH01723.1 | |
BC036051.1 | 2 | 424 | AAH36051.1 | |
BG719538.1 | 2 | 517 | ||
DA300182.1 | 2 | 420 | ||
DB049419.1 | 2 | 471 |
Target Gene Details
Entrez Gene ID: | 100529264 |
Gene Name: | RAB4B-EGLN2 readthrough (NMD candidate) |
Gene Aliases: |
RERT-lncRNA |
Location: |
Chr.19:40778219-40808441 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RAB4B-EGLN2 | NR_037791.1 | 8 | 1114 | |
AK291385.1 | 8 | 1062 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833832 | Chr.19:40754458 - 40828692 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758762 | Chr.19:40785722 - 41040163 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)