Assay Details
Target Gene Details
Entrez Gene ID: | 64333 |
Gene Name: | Rho GTPase activating protein 9 |
Gene Aliases: |
10C, RGL1 |
Location: |
Chr.12:57472255-57488814 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain | PIP4K2C MBD6 GLI1 DCTN2 DTX3 INHBE MARS DDIT3 MIR6758 MIR616 KIF5A INHBC ARHGAP9 |
nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss | PIP4K2C MBD6 GLI1 DCTN2 ARHGEF25 DTX3 MARS LOC101927583 DDIT3 MIR6758 MIR616 KIF5A ARHGAP9 |
nsv469411 | Chr.12:57450266 - 57527405 on Build GRCh38 | Loss | MIR6758 MIR616 MBD6 GLI1 INHBC INHBE MARS DDIT3 ARHGAP9 |
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | NXPH4 RDH16 RNF41 PIP4K2C PAN2 HBCBP LOC101927583 NACA ATP5B NABP2 CNPY2 SDR9C7 KIF5A R3HDM2 TIMELESS LRP1 SLC39A5 MIR1228 SNORD59A NEMP1 SLC26A10 ARHGEF25 HSD17B6 DTX3 INHBE NAB2 SPRYD4 COQ10A PRIM1 ANKRD52 TAC3 IL23A INHBC STAT6 RBMS2 SNORA105C APOF GLI1 MARS GPR182 SNORD59B ZBTB39 STAT2 STAC3 OS9 NDUFA4L2 B4GALNT1 MBD6 DCTN2 SHMT2 MIP DDIT3 PTGES3 LRP1-AS BAZ2A MIR6758 CS MIR616 MYO1A GLS2 ARHGAP9 |
nsv559035 | Chr.12:57467189 - 57476865 on Build GRCh38 | Loss | GLI1 ARHGAP9 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |