Assay Details
Target Gene Details
Entrez Gene ID: | 10480 |
Gene Name: | eukaryotic translation initiation factor 3 subunit M |
Gene Aliases: |
B5, GA17, PCID1, TANGO7, hfl-B5 |
Location: |
Chr.11:32583767-32602873 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3M | NM_001307929.1 | NP_001294858.1 | ||
NM_006360.5 | NP_006351.2 | |||
XM_011519840.2 | 2 | 193 | XP_011518142.1 | |
AF064603.1 | AAC17108.1 | |||
AK001768.1 | ||||
AK131064.1 | ||||
AK222564.1 | BAD96284.1 | |||
AK292139.1 | ||||
AK300002.1 | ||||
AK304378.1 | ||||
AK307121.1 | ||||
AK312512.1 | ||||
AV713889.1 | ||||
AY769947.1 | AAX12524.1 | |||
BC019103.2 | AAH19103.1 | |||
BC051292.1 | AAH51292.1 | |||
CR450300.1 | CAG29296.1 | |||
DB479224.1 | ||||
DQ185042.1 | ABD14422.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |