Assay Details
Target Gene Details
Entrez Gene ID: | 2026 |
Gene Name: | enolase 2 |
Gene Aliases: |
HEL-S-279, NSE |
Location: |
Chr.12:6914450-6923696 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENO2 | NM_001975.2 | 12 | 1519 | NP_001966.1 |
AK124656.1 | 11 | 1923 | ||
AK290525.1 | 12 | 1645 | ||
AK295220.1 | 10 | 1251 | ||
AK312402.1 | ||||
BC002745.2 | 12 | 1373 | AAH02745.1 | |
BT007383.1 | AAP36047.1 | |||
CR536582.1 | CAG38819.1 | |||
EU794607.1 | ||||
M22349.1 | 12 | 1371 | AAB59554.1 | |
M36768.1 | 10 | 1219 | AAA52388.1 | |
X13120.1 | CAA31512.1 | |||
X14327.1 | CAA32505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047373 | Chr.12:6902891 - 7055754 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509453 | Chr.12:6896530 - 7007852 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv557261 | Chr.12:6839304 - 6924660 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832324 | Chr.12:6780297 - 6932306 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1035811 | Chr.12:6876316 - 6994911 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)