Assay Details
Target Gene Details
Entrez Gene ID: | 26052 |
Gene Name: | dynamin 3 |
Gene Aliases: |
Dyna III |
Location: |
Chr.1:171841478-172418466 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNM3 | NM_001136127.2 | 1 | 135 | NP_001129599.1 |
NM_001278252.1 | 1 | 135 | NP_001265181.1 | |
NM_015569.4 | 1 | 135 | NP_056384.2 | |
XM_005245079.1 | 1 | 118 | XP_005245136.1 | |
XM_005245080.1 | 1 | 118 | XP_005245137.1 | |
XM_006711268.2 | 1 | 118 | XP_006711331.1 | |
XM_017000976.1 | 1 | 118 | XP_016856465.1 | |
XM_017000977.1 | 1 | 118 | XP_016856466.1 | |
XM_017000978.1 | 1 | 118 | XP_016856467.1 | |
XM_017000979.1 | 1 | 118 | XP_016856468.1 | |
XM_017000981.1 | 1 | 118 | XP_016856470.1 | |
XM_017000982.1 | 1 | 118 | XP_016856471.1 | |
XM_017000983.1 | 1 | 118 | XP_016856472.1 | |
XM_017000984.1 | 1 | 118 | XP_016856473.1 | |
XM_017000985.1 | 1 | 118 | XP_016856474.1 | |
XM_017000988.1 | 1 | 118 | XP_016856477.1 | |
XM_017000989.1 | 1 | 118 | XP_016856478.1 | |
XM_017000990.1 | 1 | 118 | XP_016856479.1 | |
XM_017000991.1 | 1 | 118 | XP_016856480.1 | |
XM_017000992.1 | 1 | 118 | XP_016856481.1 | |
AB020627.2 | 1 | 86 | ||
AK056279.1 | 1 | 115 | ||
AK307806.1 | 1 | 73 | ||
AK308992.1 | 1 | 84 | ||
AL136712.1 | 1 | 113 | CAB66647.1 | |
BC064546.1 | AAH64546.1 | |||
DC316799.1 | 1 | 132 | ||
DC334207.1 | 1 | 135 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831893 | Chr.1:171833310 - 172015860 on Build GRCh38 | Loss | DNM3-IT1 DNM3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |