Assay Details
Target Gene Details
Entrez Gene ID: | 64769 |
Gene Name: | MYST/Esa1 associated factor 6 |
Gene Aliases: |
C1orf149, CENP-28, EAF6, NY-SAR-91 |
Location: |
Chr.1:37489960-37514819 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEAF6 | NM_001270875.1 | NP_001257804.1 | ||
NM_001270876.1 | NP_001257805.1 | |||
NM_022756.5 | NP_073593.2 | |||
NR_073090.1 | ||||
NR_073091.1 | ||||
NR_073092.1 | ||||
AK021792.1 | BAB13898.1 | |||
AK225434.1 | ||||
AK225598.1 | ||||
AK308118.1 | ||||
AK309663.1 | ||||
AY007166.1 | ||||
AY211926.1 | AAO65179.1 | |||
BC016328.1 | AAH16328.1 | |||
BC056406.1 | AAH56406.1 | |||
BM840891.1 | ||||
BX386498.2 | ||||
BX538212.1 | CAD98071.1 | |||
BX640719.1 | CAE45838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519551 | Chr.1:37491775 - 37509678 on Build GRCh38 | Gain | MEAF6 MIR5581 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |