Assay Details
Target Gene Details
Entrez Gene ID: | 8409 |
Gene Name: | ubiquitously expressed prefoldin like chaperone |
Gene Aliases: |
ART-27, STAP1 |
Location: |
Chr.X:47651792-47659180 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UXT | NM_004182.3 | 5 | 383 | NP_004173.1 |
NM_153477.2 | 4 | 561 | NP_705582.1 | |
NR_045559.1 | 3 | 493 | ||
NR_045560.1 | 5 | 383 | ||
AF083241.1 | 5 | 349 | AAD39839.1 | |
AF083242.1 | 4 | 499 | AAD39840.1 | |
AF092737.1 | 5 | 364 | AAD28698.1 | |
AK223320.1 | 5 | 337 | ||
AK312072.1 | 5 | 349 | ||
BC000720.1 | 4 | 318 | AAH00720.1 | |
BC008890.1 | 4 | 318 | AAH08890.1 | |
BM471483.1 | 3 | 420 | ||
BQ681820.1 | 1 | 57 | ||
DA372561.1 | ||||
EL949569.1 | 5 | 361 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |