Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | 3 | 487 | NP_073741.2 |
AB101208.1 | 3 | 401 | ||
AK026000.1 | 3 | 394 | BAB15314.1 | |
AK292610.1 | 3 | 486 | ||
BC110910.1 | 3 | 376 | AAI10911.1 | |
BC128263.1 | 3 | 380 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | ROM1 MIR6747 MTA2 AHNAK GANAB EML3 B3GAT3 TUT1 EEF1G |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | MIR6747 MTA2 AHNAK TUT1 EEF1G |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | ROM1 MIR6747 MTA2 GANAB EML3 B3GAT3 TUT1 EEF1G |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | LRRN4CL HNRNPUL2-BSCL2 UQCC3 MIR6747 MTA2 BSCL2 AHNAK EML3 B3GAT3 SNORA57 INTS5 UBXN1 METTL12 ROM1 LBHD1 GANAB C11orf98 TUT1 EEF1G |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | LRRN4CL HNRNPUL2-BSCL2 UQCC3 MIR6747 MTA2 BSCL2 EML3 TTC9C B3GAT3 SNORA57 HNRNPUL2 INTS5 UBXN1 METTL12 ROM1 LBHD1 GNG3 GANAB C11orf98 TUT1 EEF1G |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |