Assay Details
Target Gene Details
Entrez Gene ID: | 375748 |
Gene Name: | ERCC excision repair 6 like 2 |
Gene Aliases: |
BMFS2, C9orf102, RAD26L, SR278 |
Location: |
Chr.9:95875618-96041092 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ERCC6L2 | NM_001010895.2 | NP_001010895.1 | ||
NM_020207.4 | NP_064592.2 | |||
XM_011518641.2 | XP_011516943.1 | |||
XM_011518644.2 | XP_011516946.1 | |||
XM_011518645.2 | XP_011516947.1 | |||
XM_011518646.2 | XP_011516948.1 | |||
XM_011518647.1 | XP_011516949.1 | |||
XM_017014707.1 | XP_016870196.1 | |||
AK307504.1 | ||||
AK308409.1 | ||||
AL389953.1 | CAB97543.1 | |||
BC035183.1 | AAH35183.1 | |||
BC140702.1 | ||||
DB153432.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |