Assay Details
Target Gene Details
Entrez Gene ID: | 374987 |
Gene Name: | NEXN antisense RNA 1 |
Gene Aliases: |
C1orf118 |
Location: |
Chr.1:77881348-77889539 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NEXN-AS1 | NR_103535.1 | 2 | 1172 | |
AK075118.1 | 2 | 1174 | BAC11413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2329300 | Chr.1:77887750 - 77888167 on Build GRCh38 | Deletion | NEXN-AS1 |