Assay Details
Target Gene Details
Entrez Gene ID: | 51706 |
Gene Name: | cytochrome b5 reductase 1 |
Gene Aliases: |
B5R.1, B5R1, B5R2, NQO3A2, humb5R2 |
Location: |
Chr.1:202961873-202967276 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYB5R1 | NM_016243.2 | 9 | 961 | NP_057327.2 |
AF087912.1 | 9 | 928 | AAP97209.1 | |
AF091084.1 | 4 | 386 | AAC72953.1 | |
AF093822.1 | 9 | 928 | AAP97218.1 | |
AF125533.1 | 9 | 928 | AAF17227.1 | |
AF169481.1 | 9 | 916 | AAF06147.1 | |
AK074654.1 | 9 | 942 | BAC11115.1 | |
AK075304.1 | 9 | 910 | ||
AK123705.1 | 2 | 2354 | ||
AK223416.1 | 9 | 942 | BAD97136.1 | |
AK313333.1 | ||||
AY359026.1 | 9 | 965 | AAQ89385.1 | |
BC018732.1 | 9 | 918 | AAH18732.1 | |
BC127945.1 | 6 | 648 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |