Assay Details
Target Gene Details
Entrez Gene ID: | 389362 |
Gene Name: | proteasome assembly chaperone 4 |
Gene Aliases: |
C6orf86, PAC4, bA506K6.2 |
Location: |
Chr.6:3254325-3308224 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PSMG4 | XM_011514593.2 | XP_011512895.1 | ||
AK092793.1 | 1 | 1977 | ||
AK095683.1 | ||||
AK125314.1 | 3 | 1828 |
Target Gene Details
Entrez Gene ID: | 63027 |
Gene Name: | solute carrier family 22 member 23 |
Gene Aliases: |
C6orf85 |
Location: |
Chr.6:3268973-3456574 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC22A23 | NM_001286455.1 | 10 | 4235 | NP_001273384.1 |
NM_015482.1 | 10 | 4694 | NP_056297.1 | |
NM_021945.6 | 11 | 4480 | NP_068764.3 | |
NR_104448.1 | 12 | 3951 | ||
XM_011514801.2 | 11 | 4781 | XP_011513103.1 | |
XM_017011180.1 | 11 | 5258 | XP_016866669.1 | |
XM_017011181.1 | 9 | 4007 | XP_016866670.1 | |
XM_017011182.1 | 8 | 4289 | XP_016866671.1 | |
XM_017011184.1 | 8 | 3944 | XP_016866673.1 | |
XM_017011186.1 | 11 | 4177 | XP_016866675.1 | |
AJ420525.1 | 1 | 528 | ||
AK000691.1 | 1 | 2528 | ||
AK025033.1 | 1 | 742 | BAB15054.1 | |
AK025827.1 | 1 | 439 | BAB15250.1 | |
AK172770.1 | 7 | 3790 | BAD18753.1 | |
AK225936.1 | 1 | 853 | ||
AL512737.1 | 1 | 363 | ||
BC022217.1 | 1 | 1857 | AAH22217.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic non-DGV Variation |