Assay Details
Target Gene Details
Entrez Gene ID: | 79924 |
Gene Name: | adrenomedullin 2 |
Gene Aliases: |
AM2, dJ579N16.4 |
Location: |
Chr.22:50481724-50486440 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADM2 | NM_001253845.1 | 3 | 663 | NP_001240774.1 |
XM_017028940.1 | 2 | 681 | XP_016884429.1 | |
AK024788.1 | 2 | 904 | BAB15004.1 | |
AK090635.1 | 2 | 681 | ||
AK225097.1 | 2 | 904 | ||
BX400262.2 | 2 | 540 | ||
CR456479.1 | 1 | 387 | CAG30365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3669 | Chr.22:50475511 - 50509415 on Build GRCh38 | Insertion | MIOX NCAPH2 ADM2 LMF2 |
nsv1160782 | Chr.22:50463961 - 50571524 on Build GRCh38 | Deletion | KLHDC7B CHKB-CPT1B MIOX TYMP CPT1B ODF3B NCAPH2 SCO2 ADM2 LMF2 SYCE3 SBF1 |
nsv1057656 | Chr.22:50478783 - 50595520 on Build GRCh38 | Gain | KLHDC7B CHKB-CPT1B MIOX TYMP ADM2 CHKB-AS1 CPT1B ODF3B NCAPH2 SCO2 LMF2 CHKB SYCE3 |
nsv834238 | Chr.22:50402690 - 50556965 on Build GRCh38 | Loss | PPP6R2 KLHDC7B MIOX TYMP ODF3B NCAPH2 SCO2 LOC107985556 ADM2 LMF2 SYCE3 SBF1 |
nsv471224 | Chr.22:50435189 - 50580482 on Build GRCh38 | Loss | KLHDC7B CHKB-CPT1B MIOX TYMP ADM2 SBF1 PPP6R2 CPT1B ODF3B NCAPH2 SCO2 LMF2 CHKB SYCE3 |
nsv955201 | Chr.22:50421672 - 50537371 on Build GRCh38 | Deletion | PPP6R2 MIOX TYMP ODF3B NCAPH2 SCO2 LOC107985556 ADM2 LMF2 SBF1 |
nsv829338 | Chr.22:50440870 - 50489635 on Build GRCh38 | Gain | PPP6R2 MIOX ADM2 SBF1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |