Assay Details
Target Gene Details
Entrez Gene ID: | 6357 |
Gene Name: | C-C motif chemokine ligand 13 |
Gene Aliases: |
CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1 |
Location: |
Chr.17:34356452-34358610 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL13 | NM_005408.2 | 3 | 432 | NP_005399.1 |
AJ001634.1 | 3 | 399 | CAA04888.1 | |
BC008621.1 | 3 | 395 | AAH08621.1 | |
U46767.1 | 3 | 390 | AAB38703.1 | |
U59808.1 | 3 | 432 | AAD09362.1 | |
X98306.1 | 3 | 386 | CAA66950.1 | |
Z77650.1 | 3 | 309 | CAB01111.1 | |
Z77651.1 | 2 | 250 | CAB01112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain | CCL11 LOC107987247 LOC101927239 CCL7 ASIC2 C17orf102 CCL2 CCL8 CCL13 CCL1 TMEM132E |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss | CCL11 LOC107987247 LOC101927239 CCL7 ASIC2 CCL2 CCL8 CCL13 CCL1 |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain | CCL11 LOC107987247 LOC101927239 CCL7 ASIC2 C17orf102 CCL2 CCL8 CCL13 CCL1 TMEM132E |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain | CCL11 LOC107987247 LOC101927239 CCL7 ASIC2 C17orf102 CCL2 CCL8 CCL13 CCL1 TMEM132E |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain | CCL11 LOC107987247 LOC101927239 CCL7 ASIC2 C17orf102 CCL2 CCL8 CCL13 CCL1 TMEM132E |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |