Assay Details
Target Gene Details
Entrez Gene ID: | 111 |
Gene Name: | adenylate cyclase 5 |
Gene Aliases: |
AC5, FDFM |
Location: |
Chr.3:123282296-123448632 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADCY5 | NM_001199642.1 | 19 | 2477 | NP_001186571.1 |
NM_183357.2 | 19 | 3452 | NP_899200.1 | |
XM_005247077.3 | 20 | 3614 | XP_005247134.1 | |
XM_005247078.1 | 20 | 2552 | XP_005247135.1 | |
XM_006713483.1 | 20 | 2548 | XP_006713546.1 | |
XM_006713484.1 | 20 | 2553 | XP_006713547.1 | |
XM_011512359.2 | 20 | 3412 | XP_011510661.1 | |
XM_011512360.2 | 20 | 2609 | XP_011510662.1 | |
XM_011512361.1 | 20 | 2516 | XP_011510663.1 | |
XM_017005638.1 | 20 | 2444 | XP_016861127.1 | |
XM_017005639.1 | 20 | 2444 | XP_016861128.1 | |
XM_017005640.1 | 20 | 2540 | XP_016861129.1 | |
AF497517.1 | 19 | 2353 | AAM94374.1 | |
AK093840.1 | ||||
AK124691.1 | 21 | 2964 | ||
AK294565.1 | 19 | 2645 | ||
AK302711.1 | 7 | 1023 | ||
AK303070.1 | 19 | 2478 | ||
AK316488.1 | 8 | 1065 | ||
BX647739.1 | 16 | 1954 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |